Automatic stirrer probe and paddle, Orion™
Fournisseur: Thermo Fisher Scientific
Orion™
ORIO096021EA 82.5 EUR InStock
ORIO096021 662-1553
Automatic stirrer probe and paddle, Orion™
Agitateurs Outils d'agitation
This Orion Star™ stirrer probe can be used with Orion™ Versa Star meters, Orion™ Star A210 series bench top meters and legacy Orion™ Star series bench top meters. It is designed for stirring standards and samples for better measurement accuracy without the difficulty of separate stir plate and bars.
  • Powered and controlled by the meter with five optimised stirring speeds
  • Fits in the electrode stand for easy movement in and out samples and simplified rinsing
  • Paddles can be replaced to extend the life of the stirrer probes
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité