Anti-BAX Mouse Monoclonal Antibody (Purified) [clone: 2D2]
Fournisseur: Biotium
BNUB0004-100 BNUB0004-500
BNUB0004-100EA 237 EUR InStock
BNUB0004-100 BNUB0004-500
Anti-BAX Mouse Monoclonal Antibody (Purified) [clone: 2D2]
Anticorps
Recognizes a protein of 21 kDa, identified as the Bax protein. This MAb is highly specific to Bax and shows no cross-reaction with Bcl-2 or Bcl-X protein. Bcl-2 blocks cell death following a variety of stimuli. Bax has extensive amino acid homology with Bcl-2 and it homodimerizes and forms heterodimers with Bcl-2. Overexpression of Bax accelerates apoptotic death induced by cytokine deprivation in an IL-3 dependent cell line, and Bax also counters the death repressor activity of Bcl-2.
Type: Primary
Antigen: BAX
Clonality: Monoclonal
Clone: 2D2
Conjugation: Purified
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Monkey
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité