Anti-CD4 Mouse Monoclonal Antibody (CF405S) [clone: C4/206]
Fournisseur: Biotium
BNC040206-500 BNC040206-100
BNC040206-500EA 560 EUR InStock
BNC040206-500 BNC040206-100
Anti-CD4 Mouse Monoclonal Antibody (CF405S) [clone: C4/206]
Anticorps
Recognizes a protein of 55 kDa, identified as CD4. CD4 is a membrane glycoprotein of T lymphocytes that interacts with major histocompatibility complex class II antigens and is also a receptor for the human immunodeficiency virus. This protein is expressed not only in T lymphocytes, but also in B cells, macrophages, and granulocytes. It is also expressed in specific regions of the brain. The protein functions to initiate or augment the early phase of T-cell activation, and may function as an important mediator of indirect neuronal damage in infectious and immune-mediated diseases of the central nervous system. Multiple alternatively spliced transcript variants encoding different isoforms have been identified.
Type: Primary
Antigen: CD4
Clonality: Monoclonal
Clone: C4/206
Conjugation: CF405S
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité