Anti-ERBB2 Mouse Monoclonal Antibody (CF640R) [clone: HRB2/282]
Fournisseur: Biotium
BNC400282-500 BNC400282-100
BTIUBNC400282-500EA 560 EUR InStock
BTIUBNC400282-500 BTIUBNC400282-100
Anti-ERBB2 Mouse Monoclonal Antibody (CF640R) [clone: HRB2/282]
Anticorps
Recognizes a protein of 185 kDa, which is identified as c-erbB-2/HER-2/neu. Its epitope is localized in the extracellular domain. C-erbB-2/HER-2 is a member of the EGFR family. This MAb is specific and shows minimal cross-reaction with other members of the EGFR-family. Receptors of this family are located on the plasma membrane and consist of an extracellular ligand-binding domain that is connected to a large intracellular domain by a single transmembrane sequence. c-erbB-2/HER-2 protein is over-expressed in a variety of carcinomas especially those of breast and ovary.
Type: Primary
Antigen: ERBB2
Clonality: Monoclonal
Clone: HRB2/282
Conjugation: CF640R
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité