Anti-MUC1 Mouse Monoclonal Antibody (CF568) [clone: VU-2G7]
Fournisseur: Biotium
BNC680630-100EA 192 EUR InStock
BNC680630-100 BNC680630-500
Anti-MUC1 Mouse Monoclonal Antibody (CF568) [clone: VU-2G7]
Anticorps
MAb VU-2G7 reacts with MUC1, a large transmembrane glycoprotein expressed on the ductal surface of normal glandular epithelia. The dominant epitope of VU-2G7 has not been established with epitope fingerprinting. The extracellular domain of MUC1 largely consists of a highly conserved, O-glycosylated 20 amino acids tandem repeat which can occur 30-100 times per molecule depending on the length of the allele involved. In the vast majority of human carcinomas this protein is up regulated and poorly glycosylated and appears on the cell surface in a non-polarized fashion.
Type: Primary
Antigen: MUC1
Clonality: Monoclonal
Clone: VU-2G7
Conjugation: CF568
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité