Anti-CD46 Mouse Monoclonal Antibody (CF405S) [clone: 169-1-E4.3]
Fournisseur: Biotium
BNC040930-500 BNC040930-100
BNC040930-500EA 560 EUR InStock
BNC040930-500 BNC040930-100
Anti-CD46 Mouse Monoclonal Antibody (CF405S) [clone: 169-1-E4.3]
Anticorps
Recognizes a protein of 52 kDa-58 kDa, identified as CD46 (also known as membrane cofactor protein, MCP). Its epitope is located on the four immunogenic short consensus repeat (SCR) domains at N-terminus of the protein. CD46 exists as many isoforms in a variety of tissues. It is strongly expressed on salivary gland ducts and kidney ducts, moderately on lymphocytes and endothelium, and weakly on interstitial tissues and muscle cells, but not on erythrocytes. CD46 functions as a C3b/C4b-binding glycoprotein that inhibits complement activation on host cells. It also serves as a measles virus receptor, an adherence factor for group A Streptococcus pyogenes, and a cellular pilus receptor for pathogenic Neisseria.
Type: Primary
Antigen: CD46
Clonality: Monoclonal
Clone: 169-1-E4.3
Conjugation: CF405S
Epitope:
Host: Mouse
Reactivity: Baboon, Human
Isotype: IgG2a, kappa
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité