Anti-HSP90B1 Mouse Monoclonal Antibody (CF680) [clone: 9G10.F8.2]
Fournisseur: Biotium
BNC800940-500 BNC800940-100
BTIUBNC800940-500EA 560 EUR InStock
BTIUBNC800940-500 BTIUBNC800940-100
Anti-HSP90B1 Mouse Monoclonal Antibody (CF680) [clone: 9G10.F8.2]
Anticorps
Recognizes a protein of 94 kDa, which is identified as the glucose-regulated protein 94 (grp94) and also tumor rejection antigen (gp96). Grp94 shows a high degree of sequence homology with the heat shock protein 90 (hsp90). This MAb is highly specific to grp94 and shows minimal cross-reaction with other members of the HSP90 family. Grp s are a class of proteins unresponsive to heat shock and are induced by glucose deprivation. Grp94 has been briefly studied as a prognostic factor in breast cancer.
Type: Primary
Antigen: HSP90B1
Clonality: Monoclonal
Clone: 9G10.F8.2
Conjugation: CF680
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Chicken, Cow, Dog, Guinea Pig, Hamster, Horse, Human, Monkey, Mouse, Pig, Rabbit, Rat, Sheep, Xenopus laevis
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité