Anti-CD1A Mouse Monoclonal Antibody (CF594) [clone: 66IIC7]
Fournisseur: Biotium
BNC941034-500 BNC941034-100
BNC941034-500EA 560 EUR InStock
BNC941034-500 BNC941034-100
Anti-CD1A Mouse Monoclonal Antibody (CF594) [clone: 66IIC7]
Anticorps
At least five CD1 genes (CD1a, b, c, d, and e) are identified. CD1 proteins have been demonstrated to restrict T cell response to non-peptide lipid and glycolipid antigens and play a role in non-classical antigen presentation. CD1a is a non-polymorphic MHC Class 1 related cell surface glycoprotein, expressed in association with Beta-2 microglobulin. Anti-CD1a labels Langerhans cell histiocytosis (Histiocytosis X), extranodal histiocytic sarcoma, a subset of T-lymphoblastic lymphoma/leukemia, and interdigitating dendritic cell sarcoma of the lymph node. When combined with antibodies againstMore Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité