Anti-TOX3 Mouse Monoclonal Antibody (CF640R) [clone: TOX3/1124]
Fournisseur: Biotium
BNC401124-500 BNC401124-100
BNC401124-500EA 560 EUR InStock
BNC401124-500 BNC401124-100
Anti-TOX3 Mouse Monoclonal Antibody (CF640R) [clone: TOX3/1124]
Anticorps
This antibody recognizes a 63 kDa protein, which is identified as TOX3. It contains a high mobility group (HMG)-box, which regulates Ca2 -dependent transcription in neurons through interaction with the cAMP-response-element-binding protein (CREB). TOX3 appears to be associated with breast cancer susceptibility and is expressed downstream of a cytoprotective cascade together with CITED1, a transcriptional regulator that does not bind directly to DNA. TOX3 is predominantly expressed in the brain and forms homodimers. TOX3 overexpression protects neuronal cells from cell death caused by endoplasmic reticulum stress or BAX overexpression through the induction of anti-apoptotic transcripts and repression of pro-apoptotic transcripts.
Type: Primary
Antigen: TOX3
Clonality: Monoclonal
Clone: TOX3/1124
Conjugation: CF640R
Epitope:
Host: Mouse
Isotype: IgG1 kappa1 kappa
Reactivity: Human
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité