GC columns, Rt-XLSulfur (packed and micropacked)
Fournisseur: Restek
19804 19805 19806 19807 80484-800 80484-810 80485-800 80485-810 80485-840
REST19804EA 480 EUR InStock
REST19804 REST19805 REST19806 REST19807 REST80484-800 REST80484-810 REST80485-800 REST80485-810 REST80485-840
GC columns, Rt-XLSulfur (packed and micropacked)
Colonnes pour chromatographie
These Rt-XLSulfur columns are optimised for low ppbv sulfur analyses. Interior wall and the end fittings of Rt-XLSulfur columns are Sulfinert treated, making the column as inert as PTFE.
  • Eliminate the need for PTFE tubing
  • Column and end fittings are Sulfinert treated for maximum inertness
  • Maximum temperature of 290 °C
  • Mesh size of 100/120
The packing material for Rt®-XLSulfur columns is extensively deactivated for analysis of low ppbv levels of hydrogen sulfide and methyl mercaptan. It is then treated to achieve effective separation of hydrocarbons from sulfur compounds. Extra care taken to manufacture this column ensures more accurate analyses of sulfur compounds.
Informations de commande: Columns do not include column nuts and ferrules. Optional installation kits can be ordered separately.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité