Capillary columns, Rtx®-35 amine (fused silica)
Fournisseur: Restek
11335 11338 11350 11351 11352 11353 11354 11355 11366 11369 11385
REST11335EA 599 EUR InStock
REST11335 REST11338 REST11350 REST11351 REST11352 REST11353 REST11354 REST11355 REST11366 REST11369 REST11385
Capillary columns, Rtx®-35 amine (fused silica)
Colonnes pour chromatographie
These application-specific, midpolarity phase, Crossbond 35% diphenyl/65% dimethyl polysiloxane, columns, are ideal for amines and other basic compounds, including alkylamines, diamines, triamines, ethanolamines, and nitrogen-containing heterocyclics.
  • Temperature range: stable to 220 °C
  • Chemically altered tubing surface to reduce tailing of basic compounds
  • Eliminates the need for column priming
Amine column is tested to ensure that it meets the requirements for analysing ppm levels of amines, without priming, and to ensure low bleed at maximum operating temperature. Also allows the analysis of neutral compounds and adsorptive compounds with oxygen groups susceptible to hydrogen bonding.
Note: Use base-deactivated fused silica guard columns and base-deactivated liners with Rtx-35 Amine columns.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité