NGAL (rat) ELISA kit
Fournisseur: Enzo Life Sciences
ALEXBPD-KIT-046EA 1180 EUR InStock
ALEXBPD-KIT-046 ENZOBPDKIT046
NGAL (rat) ELISA kit
Tests Tests ELISA
For the quantitative determination of NGAL in rat urine, plasma, serum, tissue extracts, or culture supernatants.
  • Ready to use pre-coated plate and individual calibrators save time and reduce errors
  • Fast results from up to 40 samples in duplicate in just 4 hours per 96 well plate
  • Absorbance is read at 450 nm
A colorimetric immunometric ELISA kit for the rapid measurement of NGAL in rat samples. Absorbance is read at 450nm.
Information de livraison: The kit contains a microwell plate coated with anti-NGAL (rat) antibody (96 wells), 5X sample diluent, NGAL rat calibrators 1 - 8, 25X wash solution concentrate, biotinylated rat NGAL antibody, HRP-streptavidin, TMB substrate and stop solution.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité