Human kallistatin ELISA kit
Fournisseur: Biorbyt
BIRBORB138068-96EA 0 EUR InStock
BIRBORB138068-96
Human kallistatin ELISA kit
Tests Tests ELISA
Human Kallistatin ELISA kit for the quantitative detection of human kallistatin in cell culture supernates, serum and plasma (heparin, EDTA).
Serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), member 4, also called Kallistatin, is a protein that in humans is encoded by the SERPINA4 gene. This gene is mapped to chromosome 14q32.13. Kallistatin is a serine proteinase inhibitor (serpin) and a heparin-binding protein. It is localized in vascular smooth muscle cells and endothelial cells of blood vessels, suggesting that it may be involved in the regulation of vascular function. It shows that kallistatin plays a role in neointima hyperplasia. The potential role of kallistatin was investigated in angiogenesis in vitro and in vivo, and presented results demonstrating a novel role of the protein in the inhibition of angiogenesis and tumour growth.
Commander maintenant
COMMANDE
SPÉCIFICATIONS
Documentation
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité