Capillary columns, Rtx®-65 (fused silica)
Fournisseur: Restek
17023 17038 17039 17054 17055
REST17023EA 803 EUR InStock
REST17023 REST17038 REST17039 REST17054 REST17055
Capillary columns, Rtx®-65 (fused silica)
Colonnes pour chromatographie
Mid-to high-polarity phase, Crossbond® diphenyl dimethyl polysiloxane, general-purpose columns for phenols, fatty acids, triglycerides.
  • Temperature range from 50 to 300 °C
  • Equivalent to USP G17 phase
  • More thermo stabile
The Rtx®-65 phase contains the highest phenyl content of any bonded stationary phase available to improve separation of aromatic compounds through increased phaseanalyte interaction. A unique polarity makes these columns ideal for a variety of analyses, from phenols to FAMEs.
As a confirmation column for EPA Method 604 phenols, an Rtx®-65 column produces a different elution order compared to the primary Rtx®-5 column. These columns elute FAMEs according to equivalent chain length, similar to bonded Carbowax® columns, but the Rtx®-65 phase does not suffer the thermal stability limitations of other polar stationary phases.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité