Bovine gamma-globulin (from Plasma)
Fournisseur: MP Biomedicals
ICNA0219147825EA 555 EUR InStock
ICNA0219147825 ICNA0219147880 ICNA0219147805
Bovine gamma-globulin (from Plasma)
Protéines et Peptides
Bovine γ-globulin is a plasma protein fraction rich in antibodies.
  • Soluble in normal saline solution (25 mg/ml, 0,9% NaCl)
It has been used as an immunological challenge in studies of mechanisms of immunotolerance. It is used to study the constituents and pH changes in protein rich hyaluronan solution which affect the biological properties of artificial articular joints. It is also used to determine immunoglobulin G in bovine colostrum and milk using direct biosensor SPR-immunoassay.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité