Bovine serum albumin (BSA), Cohn Fraction V
Fournisseur: MP Biomedicals
ImmunO™
ICNA08840533EA 393 EUR InStock
ICNA08840533 ICNA08840532
Bovine serum albumin (BSA), Cohn Fraction V
Protéines et Peptides
Albumins are a group of acidic proteins which occur plentifully in the body fluids and tissues of mammals and in some plant seeds.
  • Soluble in water
  • Viruses (blue tongue virus, vesicular stomatitis virus) were not detected
It is used to solubilise lipids, and is also used as a blocking agent in Western blots or ELISA applications. Globulin free albumins are suitable for use in applications where no other proteins should be present such as electrophoresis.
Certain conformational and primary-sequence epitopes of BSA are suspected allergens in human beef and milk allergies.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité