Bovine collagen
Fournisseur: MP Biomedicals
ICNA0219349220EA 251 EUR InStock
ICNA0219349220
Bovine collagen
Protéines et Peptides
Collagen is a group of naturally occurring proteins found in animals, especially in the flesh and connective tissues of vertebrates. It is a component of skin, bone, tendon, and other fibrous connective tissues.
It is used to form actual culture gels, and human fibroblasts cultured in these gels are able to condense the collagen to a corium-like structure by cell contraction. A highly purified collagen solution suitable for 3-D matrix formation in cell culture.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité