Injection port repair tool
Fournisseur: Restek
REST21393EA 0 EUR InStock
REST21393
Injection port repair tool
Sets d'outils
Injection port repair tool for agilent split/splitless injection ports.
  • Remove contaminants from inside the injector to achieve a better seal
  • Cleans critical inlet seal areas
  • Saves time by preventing unwanted leaks
The sealing ring at the base of a split/splitless injector allows a seal to form between the injection port and the inlet disk. This sealing ring wears over time and may become scratched or pitted, which compromises the sealing ability of the injector to the inlet disk.
Information de livraison: Includes a handle and brush adaptor, 5 medium sanding disks, 5 fine sanding disks, one 6,5 mm bore brush, and one 7 mm bore brush.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité