Pierce™ Western Blot Signal Enhancer
Fournisseur: Thermo Fisher Scientific
Pierce™
PIER21050EA 196 EUR InStock
PIER21050
Pierce™ Western Blot Signal Enhancer
Réactifs d'électrophorèse Western Blotting Reagents
Western blot signal enhancer is a membrane treatment that has been shown to increase intensity of a target protein band on a Western blot and to enable detection of a target analyte at a Level at which it could not be detected without enhancement.
  • Protein targets can be detected at lower levels
  • Enhances antibody binding to immobilised antigens
  • Works with any substrate, enhancing both chemiluminescence and colorimetric detection
  • Works with any membrane
  • Ready-to-use and can be stored at room temperature
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité