Liquid tight chemical suit, Microchem® 4000 Apollo, model 126
Fournisseur: ANSELL HEALTH CARE
GR40T-00126-03 GR40T-00126-04 GR40T-00126-05 GR40T-00126-06
ANSEGR40T-00126-03EA 0 EUR InStock
ANSEGR40T-00126-03 ANSEGR40T-00126-04 ANSEGR40T-00126-05 ANSEGR40T-00126-06
Liquid tight chemical suit, Microchem® 4000 Apollo, model 126
Tenue de travail
This fully encapsulated liquid tight chemical suit is designed for use in conjunction with self contained breathing apparatus (SCBA). Developed with the UK Fire & Rescue services.
  • Rear entry double zip system
  • Rear mounted BA (breathing apparatus) pouch – universal fit with most BA
  • Attached socks with boot overflap and attached Ansell Barrier® gloves
  • Exhalation valves, clear face visor
  • Ultrasonically welded and taped seams
  • Bat-wing design enables air gauge checking within the suit
  • Chest strap for DSU (Distress Signal Unit), adjustable internal support braces
Typical applications include: Chemicals, oil and petrolchemicals, pharmaceutical, agriculture, sewage purification installations, industrial, tank cleaning and emergency services (HAZMAT, CBRN). Also suitable for use in level-B applications (according to US Environmental Protection Agency (EPA) & NFPA guidelines). Suitable for Ex-Zones.
Certifications: Type 3/4, EN 14126; Cat. III
Suitable for Ex-Zones
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité