Apron with sleeves, MICROCHEM® 3000, model 214
Fournisseur: Ansell
Microchem® 3000
114-9515EA 731 EUR InStock
114-9515 114-9514 114-9513 114-9512 114-9511
Apron with sleeves, MICROCHEM® 3000, model 214
Vêtements à usage unique
This apron is made from an external film barrier layer with two non woven layers underneath. The fabric provides a strong liquid and particle barrier.
  • Rear velcro fastening
  • Elasticated wrists, welded seams
  • Eltrasonically welded elastication to waist
  • Multi-layer barrier fabric is effective against numerous chemicals
  • Highly visible: Bright yellow for improved worker safety
  • Lightweight yet durable
Typical applications include: Chemicals, oil and petrochemicals, pharmaceutical, food industry (caustic clean downs), sewage purification installations, industrial and tank cleaning, mining.
Certifications: Type 3-B/4-B/5-B, EN 14126, EN 1073-2, EN 1149-5.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité