Sleeve protectors, MICROCHEM® 3000, model 600
Fournisseur: Ansell
Microchem® 3000
114-9517EA 636 EUR InStock
114-9517
Sleeve protectors, MICROCHEM® 3000, model 600
Vêtements à usage unique
These sleeve protectors are made from a multi-layer fabric (external film barrier layer, non woven inner layer, core high barrier film layer) that provides an extremely effective barrier against both inorganic chemicals and biological hazards.
  • Elasticated at both ends
  • Welded seams
  • Lightweight and comfortable
  • Excellent liquid penetration resistance and barrier to fine particulates
  • Silicone free: Critical in paint spraying applications
  • Anti-static: Tested according to EN 1149-5
Typical applications include: Chemicals, oil and petrochemicals, pharmaceutical, food industry (caustic clean downs), sewage purification installations, industrial and tank cleaning, and mining.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité