Bovine native collagen (from Achilles tendon)
Fournisseur: Abnova
ABNOP5215EA 132 EUR InStock
ABNOP5215
Bovine native collagen (from Achilles tendon)
Protéines et Peptides
Collagen is an inert, rigid protein found predominantly in skin, ligaments, bones and teeth. It is a lyophilised, shredded insoluble preparation, used as a substrate for collagenase.
Its most distinctive characteristic in its role as a transmitter of mechanical force is its inelasticity. It is stable for at least three months.
Commander maintenant
COMMANDE
SPÉCIFICATIONS
Documentation
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité