Anti-TrkB (phospho-Ser478) Rabbit Polyclonal Antibody
Fournisseur: Biosensis
BSENR-1718-50EA 302 EUR InStock
BSENR-1718-50
Anti-TrkB (phospho-Ser478) Rabbit Polyclonal Antibody
Anticorps
This antibody has been shown to be specific for TrkB phosphorylated on serine 478 by phospho-peptide absorption dot blots, and on cell lysates from cell lines induced with retinoic acid and BDNF. Antibody detects a clear band in retinoic acid (RA) and BDNF-treated NSC34 cell lysates at 140 kDa only, indicating that the phosphorylated TrkB receptor is being detected.
  • Recommeded 0,5 to 2 µg/ml for western blotting and 1 to 5 µg/ml for immunohistochemistry
  • TBS is preferred for buffer preparation
Cross-reactivity: Human and mouse, other species not yet tested but it is predicted to react with rat and chicken TrkB-pS478 based on amino acid homology at the site.
Type: Primary
Antigen: TrkB phospho S478
Clonality: polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Commander maintenant
COMMANDE
SPÉCIFICATIONS
Documentation
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité