Real-Time PCR kit for detection of mycoplasma, Microsart® AMP Mycoplasma
Fournisseur: Sartorius
SARTSMB95-1002EA 2640 EUR InStock
SARTSMB95-1002 SARTSMB95-1001
Real-Time PCR kit for detection of mycoplasma, Microsart® AMP Mycoplasma
AMP Mycoplasma Kit is used for direct detection of Mollicutes (Mycoplasma, Acholeplasma, Spiroplasma) in cell cultures, cell culture media components and derived biologicals.
  • Minimises testing time from weeks down to hours
  • Convenient and simple to use
  • For use with all common qPCR cyclers able to detect FAM ™- and ROX™-labeled probes
  • Testing volume from 200 µl up to 18 ml ensures highest sensitivity
Rapid methods for the detection of mycoplasma contamination are essential for improving monitoring of the production process of biologicals. The Microsart® AMP Mycoplasma can be easily implemented because no specific hardware is needed. The kit can be used with any type of real-time PCR cycler that is able to detect the fluorescence dyes FAM™ and ROX™. The total testing procedure takes only 3 hours.
The kit contains all essential components in a function tested ready-to-use master mix, including the polymerase.
Certifications: Validated according to EP 2.6.7.
Information de livraison: Kit contains Mycoplasma mix, rehydration buffer, positive control DNA, internal control DNA, PCR grade water.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité