Human recombinant PEF (1-284) (from E. coli)
Fournisseur: ProSci
PRSIXW-RP3186EA 690 EUR InStock
PRSIXW-RP3186
Human recombinant PEF (1-284) (from E. coli)
Protéines et Peptides
Peflin has a longer N-terminal hydrophobic domain than any other member of the family, and it contains nine nonapeptide (A/PPGGPYGGP) repeats. Western blot analysis demonstrated that peflin (30 kDa) was expressed in various nonadherent and adherent cultured human cell lines, including Jurkat, HL60, HeLa, and HT1080. Peflin may play basic roles in Ca(2+) signaling irrespective of cell types.
  • This recombinant protein can be used for WB, ELISA, MS.
10 mM Tris, pH 8.0, 0.002% NaN, 3mM NaCl, 2.5mM
Store in working aliquots at -80°C. Avoid freeze/thaw cycles.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité