Human recombinant IL15 (from E. coli)
Fournisseur: ProSci
PRSI40-558EA 412 EUR InStock
PRSI40-558
Human recombinant IL15 (from E. coli)
Protéines et Peptides
IL-15 is an immunomodulating cytokine that stimulates the proliferation of T lymphocytes and shares many biological properties with IL-2. IL-15 exerts its biological activities primarily on T cells. It is also essential in the development, survival and activation of NK cells. Increased expression of IL-15 has been implicated with rheumatoid arthritis, inflammatory bowl disease and diseases affiliated with retroviruses HIV and HTLV-I. Human IL-15 is biologically active on mouse cells as measured by the dose-dependent stimulation of the proliferation of mouse CTLL cells.
  • Recombinant human (40-260) IL-15 is a 12.9 kDa protein consisting of 114 amino acid residues. Recombinant human IL-15 (40-558) is a 12.9 kDa protein consisting of 115 amino acid residues. Recombinant murine IL-15 is a 13.3 kDa protein consisting of 115 amino acid residues. Recombinant rat IL-15 is a 13.5 kDa protein consisting of 115 amino acid residues.
Store in working aliquots at -20°C. Avoid freeze/thaw cycles.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité