Human recombinant IFNG (from E. coli)
Fournisseur: ProSci
PRSI40-557EA 412 EUR InStock
PRSI40-557
Human recombinant IFNG (from E. coli)
Protéines et Peptides
IFN-γ is an acid-labile interferon produced by CD4 and CD8 T lymphocytes as well as activated NK cells. IFN-γ receptors are present in most immune cells, which respond to IFN-γ signaling by increasing the surface expression of class I MHC proteins. This promotes the presentation of antigen to T-helper (CD4+) cells. IFN-γ signaling in antigen-presenting cells and antigen-recognizing B and T lymphocytes regulate the antigen-specific phases of the immune response. Additionally, IFN-γ stimulates a number of lymphoid cell functions including the anti-microbial and anti-tumor responses of macrophages, NK cells, and neutrophils. Human IFN-γ species-specific and is biologically active only in human and primate cells. Recombinant human IFN-γ is a 16.7 kDa protein containing 143 amino acid residues.Manufactured using all non-animal reagents.
  • Store in working aliquots at -20°C. Avoid freeze/thaw cycles.
Commander maintenant
COMMANDE
SPÉCIFICATIONS
Documentation
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité