Human recombinant TGF alpha (from E. coli)
Fournisseur: ProSci
PRSI40-573EA 412 EUR InStock
PRSI40-573
Human recombinant TGF alpha (from E. coli)
Protéines et Peptides
TGF-α is an EGF-related polypeptide growth factor that signals through the EGF receptor, and stimulates the proliferation of a wide range of epidermal and epithelial cells. It is produced by monocytes, keratinocytes, and various tumor cells. TGF-α induces transformation anchorage independence in cultured cells. Human, murine and rat TGF-α are cross-species reactive. Recombinant human TGF-α is a 50 amino acid polypeptide (5.5 kDa) which shares approximately 40% sequence homology with EGF, including 6 conserved cysteine residues, which form 3 intramolecular disulfide bonds.
  • Store in working aliquots at -20°C. Avoid freeze/thaw cycles.
Commander maintenant
COMMANDE
SPÉCIFICATIONS
Documentation
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité