Rat recombinant thrombopoietin (from E. coli)
Fournisseur: ProSci
PRSI40-537EA 412 EUR InStock
PRSI40-537
Rat recombinant thrombopoietin (from E. coli)
Protéines et Peptides
TPO is a lineage specific growth factor, produced in the liver, kidney and skeletal muscle. It stimulates the proliferation and maturation of megakaryocytes, and promotes increased circulating levels of platelets in vivo. TPO signals through the c-mpl receptor and acts as an important regulator of circulating platelets. Human and murine TPO exhibits cross-species reactivity. Recombinant rat TPO is a fully biologically active 174 amino acid polypeptide (18.7 kDa), which contains the erythropoietin-like domain of the full length TPO protein.
  • Store in working aliquots at -20°C. Avoid freeze/thaw cycles.
Commander maintenant
COMMANDE
SPÉCIFICATIONS
Documentation
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité