Human recombinant ANGPTL3 (from HEK293 cells)
Fournisseur: ProSci
PRSI96-027EA 612 EUR InStock
PRSI96-027
Human recombinant ANGPTL3 (from HEK293 cells)
Protéines et Peptides
Angiopoietin-like protein 3 (ANGPTL3) is also known as Angiopoietin-related protein 3, Angiopoietin-5 (ANGPT5 / ANG-5), is a member of the angiopoietin-like family of secreted factors. ANGPTL3 / ANGPT5 is predominantly expressed in the liver, and has the characteristic structure of angiopoietins, consisting of a signal peptide, N-terminal coiled-coil domain and the C-terminal fibrinogen (FBN)-like domain. The FBN-like domain in angiopoietin-like 3 protein was shown to bind alpha-5/beta-3 integrins, and this binding induced endothelial cell adhesion and migration. This protein may also play a role in the regulation of angiogenesis. Angptl3 also acts as dual inhibitor of lipoprotein lipase (LPL) and endothelial lipase (EL), and increases plasma triglyceride and HDL cholesterol in rodents. ANGPTL3 inhibit endothelial lipase to catalyze HDL-phospholipid and increase HDL-PL levels. Circulating PL-riched HDL particles have high cholesterol efflux abilities.
Commander maintenant
COMMANDE
SPÉCIFICATIONS
Documentation
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité