Human recombinant Azurocidin (from HEK293 cells)
Fournisseur: ProSci
PRSI96-038EA 553 EUR InStock
PRSI96-038
Human recombinant Azurocidin (from HEK293 cells)
Protéines et Peptides
Azurocidin (AZU1) is also known as Heparin-binding protein (HBP), Cationic antimicrobial protein CAP37, is a neutrophil granule-derived antibacterial and monocyte- and fibroblast-specific chemotactic glycoprotein. which belongs to the peptidase S1 family and elastase subfamily. AZU1 / HBP contains 1 peptidase S1 domain. AZU1 binds heparin. The cytotoxic action of AZU1 is limited to many species of Gram-negative bacteria; this specificity may be explained by a strong affinity of the very basic N-terminal half for the negatively charged lipopolysaccharides that are unique to the Gram-negative bacterial outer envelope. AZU1 may play a role in mediating recruitment of monocytes in the second wave of inflammation.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité