Human recombinant Cystatin D (from HEK293 cells)
Fournisseur: ProSci
PRSI96-209EA 612 EUR InStock
PRSI96-209
Human recombinant Cystatin D (from HEK293 cells)
Protéines et Peptides
Cystatin-5 (CST5) is also known as Cystatin-D, is a secreted thermostable protein, which belongs to the cystatin family. CST5 is a cysteine proteinase inhibitor that possibly plays a protective role against proteinases present in the oral cavity. The order of preference for inhibition is cathepsin S > cathepsin H > cathepsin L > cathepsin B. CST5 / cystatin-D may play a protective role against proteinases present in the oral cavity.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité