Human recombinant Bone Proteoglycan II (from HEK293 cells)
Fournisseur: ProSci
PRSI96-234EA 553 EUR InStock
PRSI96-234
Human recombinant Bone Proteoglycan II (from HEK293 cells)
Protéines et Peptides
Decorin is also known as bone proteoglycan II, PGS2, SLRR1B, DCN, DSPG2 and PG40, is a secreted chondroitin /dermatan sulfate proteoglycan in the family of small leucine-rich proteoglycans (SLRPs). Decorin is a small cellular or pericellular matrix proteoglycan and is closely related in structure to biglycan protein. Decorin and biglycan are thought to be the result of a gene duplication. This protein is a component of connective tissue, binds to type I collagen fibrils, and plays a role in matrix assembly. Decorin appears to influence fibrillogenesis, and also interacts with fibronectin, thrombospondin, the complement component C1q, epidermal growth factor receptor (EGFR) and transforming growth factor-beta (TGF-beta). Defects in DCN are the cause of congenital stromal corneal dystrophy (CSCD).
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité