Human recombinant lipopolysaccharide (from HEK293 cells)
Fournisseur: ProSci
PRSI96-506EA 612 EUR InStock
PRSI96-506
Human recombinant lipopolysaccharide (from HEK293 cells)
Protéines et Peptides
Lipopolysaccharide-binding protein (LBP),a member of the BPI/LBP/Plunc superfamily, BPI/LBP family, is detected in blood serum. LBP plays a role in the innate immune response. Alao,LBP can bind to the lipid A moiety of bacterial lipopolysaccharides (LPS), a glycolipid present in the outer membrane of all Gram-negative bacteria, and act as an affinity enhancer for CD14, facilitating its association with LPS. Furthermore, LBP is able to promote the release of cytokines in response to bacterial lipopolysaccharide.
Commander maintenant
COMMANDE
SPÉCIFICATIONS
Documentation
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité