Human recombinant LYPD3 (from HEK293 cells)
Fournisseur: ProSci
PRSI96-528EA 553 EUR InStock
PRSI96-528
Human recombinant LYPD3 (from HEK293 cells)
Protéines et Peptides
Ly6/PLAUR domain-containing protein 3 (LYPD3) is also known as MIG-C4 ,C4.4A, GPI-anchored metastasis-associated protein C4.4A homolog and Matrigel-induced gene C4 protein,which is a cell membrane protein containing two UPAR/Ly6 domains. LYPD3 is expressed in placenta, skin and urothelium. LYPD3 is found in the majority of primary and metastatic transitional cell carcinomas (TCCs) and as well in breast cancer tissues, but not in adjacent normal tissues. LYPD3 play a role in supporting cell migration and it is not only probably involved in urothelial cell-matrix interactions, but also in tumor progression.
Commander maintenant
COMMANDE
SPÉCIFICATIONS
Documentation
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité