Human recombinant Sectm1 (from HEK293 cells)
Fournisseur: ProSci
PRSI96-682EA 525 EUR InStock
PRSI96-682
Human recombinant Sectm1 (from HEK293 cells)
Protéines et Peptides
Secreted and transmembrane protein 1 (SECTM1) is also known as Protein K-12, which is belongs to the SECTM family, which is Inducted by IFNG/IFN-gamma (at protein level). SECTM1 is detected at the highest levels in peripheral blood leukocytes and breast cancer cell lines. SECTM1 may be involved in thymocyte signaling.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité