Human recombinant Stathmin (from E. coli)
Fournisseur: ProSci
PRSI96-705EA 454 EUR InStock
PRSI96-705
Human recombinant Stathmin (from E. coli)
Protéines et Peptides
Stathmin (STMN1), a member of the stathmin family, is also known as Leukemia-associated phosphoprotein p18, Oncoprotein 18 (Op18), Phosphoprotein p19 (pp19), Protein Pr22 and pp17, which contains one SLD (stathmin-like) domain. The function of STMN1 as an important regulatory protein of microtubule dynamics has been well-characterized. Stathmin (STMN1) interacts with two molecules of dimeric α,β-tubulin to form a tight ternary complex called the T2S complex.One mole of STMN1 binds to two moles of tubulin dimers through the stathmin-like domain (SLD).When STMN1 sequesters tubulin into the T2S complex, tubulin becomes non-polymerizable. Without tubulin polymerisation, there is no microtubule assembly.STMN1 also promotes microtubule disassembly by acting directly on the microtubule ends.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité