Human recombinant DR6 (from HEK293 cells)
Fournisseur: ProSci
PRSI96-730EA 584 EUR InStock
PRSI96-730
Human recombinant DR6 (from HEK293 cells)
Protéines et Peptides
Tumor necrosis factor receptor superfamily member 21 (TNFRSF21) is also known as death receptor 6 (DR6), which is a member of the TNF-receptor superfamily. TNFRSF21 contains one death domain and four TNFR-Cys repeats. TNFRSF21 / DR6 has been shown to activate NF-κB and MAPK8/JNK, and induce cell apoptosis. Through its death domain, this receptor interacts with TRADD protein, which is known to serve as an adaptor that mediates signal transduction of TNF-receptors.
Commander maintenant
COMMANDE
SPÉCIFICATIONS
Documentation
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité