Human recombinant FN14 (from HEK293 cells)
Fournisseur: ProSci
PRSI96-735EA 553 EUR InStock
PRSI96-735
Human recombinant FN14 (from HEK293 cells)
Protéines et Peptides
Tumor necrosis factor receptor superfamily member 12A (TNFRSF12A) is also known as Fibroblast growth factor-inducible immediate-early response protein 14 (FN14 or FGF-inducible 14), Tweak-receptor, CD antigen CD266. TNFRSF12A contains one TNFR-Cys repeat. TNFRSF12A is highly expressed in heart, placenta and kidney. TNFRSF12A is receptor for TNFSF12/TWEAK, and is weak inducer of apoptosis in some cell types. TNFRSF12A / FN14 promotes angiogenesis and the proliferation of endothelial cells.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité