Human recombinant LIF (from E. coli)
Fournisseur: ProSci
PRSI91-014EA 584 EUR InStock
PRSI91-014
Human recombinant LIF (from E. coli)
Protéines et Peptides
Leukemia Inhibitory Factor (LIF) is a lymphoid factor that promotes long-term maintenance of embryonic stem cells by suppressing spontaneous differentiation. LIF has a number of other activities including cholinergic neuron differentiation, control of stem cell pluripotency, bone and fat metabolism, mitogenesis of certain factor dependent cell lines and promotion of megakaryocyte production in vivo. Human and murine mature LIF exhibit a 78% sequence identity at the amino acid level. Human LIF is equally active on human and mouse cells. Murine LIF is approximately 1000 fold less active on human cells than human LIF.
  • This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité