Human recombinant FGF1 (from E. coli)
Fournisseur: ProSci
PRSI91-039EA 365 EUR InStock
PRSI91-039
Human recombinant FGF1 (from E. coli)
Protéines et Peptides
FGF acidic, also known as ECGF, FGF-1and HBGF-1, is a non-glycosylated heparin binding growth factor that is expressed in the brain, kidney, retina, smooth muscle cells, bone matrix, osteoblasts, astrocytes and endothelial cells. It is a mitogenic peptide that is produced by multiple cell types and stimulates the proliferation of cells of mesodermal, ectodermal, and endodermal origin. Its association with heparan sulfate is a prerequisite for activation of FGF receptors. Internalized FGF acidic migrates to the nucleus where it is phosphorylated by nuclear PKC delta, exported to the cytosol, dephosphorylated, and degraded. Intracellular FGF acidic inhibits p53 activity and proapoptotic signaling.
  • This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité