Human recombinant Myelin oligodendrocyte glycoprotein (from E. coli)
Fournisseur: ProSci
PRSI91-041EA 471 EUR InStock
PRSI91-041
Human recombinant Myelin oligodendrocyte glycoprotein (from E. coli)
Protéines et Peptides
Myelin Oligodendrocyte Glycoprotein (MOG) is a transmembrane protein, which is expressed exclusively in the CNS. MOG contains a single Ig-domain exposed to the extracellular space that allows autoantibodies easy access. MOG protein has been identified as a crucial autoantigen for multiple sclerosis in humans. MOG is capable to produce a demyelinating multiple sclerosis-like diseases in experimental animals, namely experimental autoimmune encephalomyelitis (EAE), in rodents and monkeys.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité