Human recombinant CXCL7 (from E. coli)
Fournisseur: ProSci
PRSI91-045EA 761 EUR InStock
PRSI91-045
Human recombinant CXCL7 (from E. coli)
Protéines et Peptides
Human Chemokine (C-X-C motif) Ligand 7 (CXCL7), also known as neutrophil activating peptide 2 (NAP-2), is a member of the CXC chemokines containing an ELR domain (Glu-Leu-Arg tripeptide motif). Similar to other ELR domain containing CXC chemokines, such as IL-8 and the GRO proteins, CXCL7 binds CXCR2, chemoattracts and activates neutrophils. CXCL7, Connective Tissue Activating Protein III (CTAPIII) and beta thrombogulin ( beta TG), are proteolytically processed carboxylterminal fragments of platelet basic protein (PBP) which is found in the alphagranules of human platelets. Although CTAPIII, beta TG, and PBP represent amino-terminal extended variants of NAP2 and possess the same CXC chemokine domains, these proteins do not exhibit CXCL7/NAP2 activity. CXCL7 induces cell migration through the G-protein-linked receptor CXCR-2.
  • This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité