Human recombinant Neurotrophin 3 (from E. coli)
Fournisseur: ProSci
PRSI91-065EA 761 EUR InStock
PRSI91-065
Human recombinant Neurotrophin 3 (from E. coli)
Protéines et Peptides
Neurotrophin-3 (NT-3) is a member of the NGF family of neurotrophic factors and is structurally related to beta -NGF, BDNF and NT-4. The NT3 cDNA encodes a 257 amino acid residue precursor protein with a signal peptide and a proprotein that are cleaved to yield the 119 amino acid residue mature NT3.The amino acid sequences of mature human, murine and rat NT-3 are identical. NT-3 selectively promotes the differentiation and survival of specific neuronal subpopulations in both the central as well as the peripheral nervous systems.
  • This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité