Rabbit Recombinant TNF alpha (from E. coli)
Fournisseur: ProSci
PRSI91-068EA 655 EUR InStock
PRSI91-068
Rabbit Recombinant TNF alpha (from E. coli)
Protéines et Peptides
Tumor necrosis factor alpha (TNF alpha) is the prototypic ligand of the TNF superfamily. TNF alpha forms a homotrimer and functions by activating two types of receptors TNF-R1 (TNF receptor type 1,p55R) and TNF-R2 (TNF receptor type 2,p75R). TNF alpha is a pleiotropic cytokine that is capable to promote inflammation, to induce apoptotic cell death, and to inhibit tumorigenesis and viral replication. TNF alpha is a potent lymphoid factor that exerts cytotoxic effects on a wide range of tumor cells and certain other target cells.
  • This recombinant protein can be used for biological assays. For research use only.
Commander maintenant
COMMANDE
SPÉCIFICATIONS
Documentation
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité