Human recombinant APT-1 (from E. coli)
Fournisseur: ProSci
PRSI91-080EA 584 EUR InStock
PRSI91-080
Human recombinant APT-1 (from E. coli)
Protéines et Peptides
Acyl-Protein Thioesterase 1 (APT-1) is lysophospholipase that belongs to the AB hydrolase 2 family. APT-1 performs on biological membranes to regulate the multifunctional lysophospholipids. It hydrolyzes fatty acids from S-acylated cysteine residues in proteins such as trimeric G alpha proteins or HRAS; in addition, it also has depalmitoylating activity and low lysophospholipase activity.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: N-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité