Human recombinant Cytoglobin (from E. coli)
Fournisseur: ProSci
PRSI91-107EA 584 EUR InStock
PRSI91-107
Human recombinant Cytoglobin (from E. coli)
Protéines et Peptides
Cytoglobin is a ubiquitously globin protein that belongs to the globin family. The highest expressed in heart, stomach, bladder and small intestine. CYGB acts a protector under conditions of oxidative stress. CYGB may be involved in intracellular oxygen storage or transfer, modulates oxygen and nitric oxide metabolism or scavenging free radicals within a cell.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité