Human recombinant FABP7 (from E. coli)
Fournisseur: ProSci
PRSI91-121EA 584 EUR InStock
PRSI91-121
Human recombinant FABP7 (from E. coli)
Protéines et Peptides
Fatty Acid-Binding Protein 7 (FABP7) is a cytoplasm protein that belongs to the Fatty-acid Binding Protein (FABP) family of calycin superfamily. Fatty acid binding proteins are a family of small, highly conserved, cytoplasmic proteins that bind long-chain fatty acids. FABP7 is predominately expressed in brain and neural tissues. FABP7 is involved in fatty acid uptake and intracellular transport and is important in brain development. FABP7 plays a critical role in the transport of a so far unknown hydrophobic ligand with potential morphogenic activity during CNS development. FABP7 is required for the establishment of the radial glial fiber system in developing brain, a system that is necessary for the migration of immature neurons to establish cortical layers.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: N-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité