Human Recombinant Grancalcin (from E. coli)
Fournisseur: ProSci
PRSI91-126EA 584 EUR InStock
PRSI91-126
Human Recombinant Grancalcin (from E. coli)
Protéines et Peptides
Grancalcin (GCA) is a member of the penta EF hand subfamily which includes sorcin, calpain and ALG2. Grancalcin is highly expressed bone marrow and also can detected in neutrophils and macrophages. Grancalcin interacts with L-plastin which known to have actin bundling activity. It indicates that Grancalcin may play an important role in the adhesion of neutrophils to fibronectin. Furthermore, Grancalcin localization is dependent upon calcium and magnesium. It associates with both the granule and membrane fractions, which suggested a role for grancalcin in granule-membrane fusion and degranulation.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: N-GST tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité